accessibility menu, dialog, popup

UserName

NH2SNHF

CAS RN 656-32-6

IUPAC Name: (2-fluorophenyl)thiourea
Synonyms: 2-fluorophenylthiourea
Molecular Weight (g/mol): 170.21
Molecular Formula: C7H7FN2S
InChi Key: WYVZQQOFMQRNPF-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=CC=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results