Learn More
2-Fluorophenyl isothiocyanate, 97%
CAS: 38985-64-7 | C7H4FNS | 153.17 g/mol
$83.14 - $225.96
Chemical Identifiers
| CAS | 38985-64-7 |
|---|---|
| Molecular Formula | C7H4FNS |
| Molecular Weight (g/mol) | 153.17 |
| MDL Number | MFCD00004800 |
| InChI Key | OAGDRIUTLPDSMJ-UHFFFAOYSA-N |
| Synonym | 2-fluorophenyl isothiocyanate, o-fluorophenyl isothiocyanate, benzene, 1-fluoro-2-isothiocyanato, 2-fluorophenylisothiocyanate, 1-fluoro-2-isothiocyanato-benzene, isothiocyanic acid 2-fluorophenyl ester, benzene,1-fluoro-2-isothiocyanato, o-fluoro phenyl isothiocyanate, 2-fluorobenzenisothiocyanate, timtec-bb sbb006546 |
| PubChem CID | 97520 |
| IUPAC Name | 1-fluoro-2-isothiocyanatobenzene |
| SMILES | FC1=CC=CC=C1N=C=S |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
AAA1831406
|
Thermo Scientific Chemicals
A1831406 |
5 g |
Each for $83.14
|
|
|||||
|
AAA1831414
|
Thermo Scientific Chemicals
A1831414 |
25 g |
Each for $225.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 38985-64-7 | |
| 153.17 | |
| OAGDRIUTLPDSMJ-UHFFFAOYSA-N | |
| 97520 | |
| FC1=CC=CC=C1N=C=S |
| C7H4FNS | |
| MFCD00004800 | |
| 2-fluorophenyl isothiocyanate, o-fluorophenyl isothiocyanate, benzene, 1-fluoro-2-isothiocyanato, 2-fluorophenylisothiocyanate, 1-fluoro-2-isothiocyanato-benzene, isothiocyanic acid 2-fluorophenyl ester, benzene,1-fluoro-2-isothiocyanato, o-fluoro phenyl isothiocyanate, 2-fluorobenzenisothiocyanate, timtec-bb sbb006546 | |
| 1-fluoro-2-isothiocyanatobenzene |
Specifications
| 38985-64-7 | |
| 103°C to 104°C (12 mmHg) | |
| C7H4FNS | |
| MFCD00004800 | |
| UN2922 | |
| Moisture sensitive | |
| OAGDRIUTLPDSMJ-UHFFFAOYSA-N | |
| 1-fluoro-2-isothiocyanatobenzene | |
| 97520 | |
| 97% |
| 1.248 | |
| 97°C (206°F) | |
| 1.6254 | |
| 5 g | |
| 4176581 | |
| 2-fluorophenyl isothiocyanate, o-fluorophenyl isothiocyanate, benzene, 1-fluoro-2-isothiocyanato, 2-fluorophenylisothiocyanate, 1-fluoro-2-isothiocyanato-benzene, isothiocyanic acid 2-fluorophenyl ester, benzene,1-fluoro-2-isothiocyanato, o-fluoro phenyl isothiocyanate, 2-fluorobenzenisothiocyanate, timtec-bb sbb006546 | |
| FC1=CC=CC=C1N=C=S | |
| 153.17 | |
| 153.18 | |
| 2-Fluorophenyl isothiocyanate |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 254-237-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only