CAS RN 38985-64-7
IUPAC Name:
1-fluoro-2-isothiocyanatobenzene
Synonyms:
1-fluoro-2-isothiocyanatobenzene
Molecular Weight (g/mol):
153.17
Molecular Formula:
C7H4FNS
InChi Key:
OAGDRIUTLPDSMJ-UHFFFAOYSA-N
SMILES:
FC1=CC=CC=C1N=C=S
1
–
2
of
2
results
1
Promotions Available
2-Fluorophenyl isothiocyanate, 97%
2-Fluorophenyl isothiocyanate, 97%
CAS: 38985-64-7 Molecular Formula: C7H4FNS Molecular Weight (g/mol): 153.17 MDL Number: MFCD00004800 InChI Key: OAGDRIUTLPDSMJ-UHFFFAOYSA-N Synonym: 2-fluorophenyl isothiocyanate,o-fluorophenyl isothiocyanate,benzene, 1-fluoro-2-isothiocyanato,2-fluorophenylisothiocyanate,1-fluoro-2-isothiocyanato-benzene,isothiocyanic acid 2-fluorophenyl ester,benzene,1-fluoro-2-isothiocyanato,o-fluoro phenyl isothiocyanate,2-fluorobenzenisothiocyanate,timtec-bb sbb006546 PubChem CID: 97520 IUPAC Name: 1-fluoro-2-isothiocyanatobenzene SMILES: FC1=CC=CC=C1N=C=S