missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenyl isothiocyanate, 97%

Catalog No. AAA1831406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1831406 5 g
AAA1831414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1831406 Supplier Thermo Scientific Chemicals Supplier No. A1831406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38985-64-7 | C7H4FNS | 153.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38985-64-7
Molecular Formula C7H4FNS
Molecular Weight (g/mol) 153.17
MDL Number MFCD00004800
InChI Key OAGDRIUTLPDSMJ-UHFFFAOYSA-N
Synonym 2-fluorophenyl isothiocyanate, o-fluorophenyl isothiocyanate, benzene, 1-fluoro-2-isothiocyanato, 2-fluorophenylisothiocyanate, 1-fluoro-2-isothiocyanato-benzene, isothiocyanic acid 2-fluorophenyl ester, benzene,1-fluoro-2-isothiocyanato, o-fluoro phenyl isothiocyanate, 2-fluorobenzenisothiocyanate, timtec-bb sbb006546
PubChem CID 97520
IUPAC Name 1-fluoro-2-isothiocyanatobenzene
SMILES FC1=CC=CC=C1N=C=S

Specifications

Density 1.248
Boiling Point 103°C to 104°C (12 mmHg)
Flash Point 97°C (206°F)
Refractive Index 1.6254
Quantity 5 g
UN Number UN2922
Beilstein 4176581
Sensitivity Moisture sensitive
Formula Weight 153.18
Percent Purity 97%
Chemical Name or Material 2-Fluorophenyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 254-237-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only