missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenol 98.0+%, TCI America™

Catalog No. p-7129506
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
F015025G 25 g
F0150250G 250 g
2 options

Catalog No. F015025G

Supplier: TCI America F015025G

Only null left

Chemical Identifiers

CAS 367-12-4
Molecular Formula C6H5FO
Molecular Weight (g/mol) 112.10
MDL Number MFCD00002155
InChI Key HFHFGHLXUCOHLN-UHFFFAOYSA-N
Synonym o-fluorophenol, phenol, 2-fluoro, phenol, o-fluoro, 1-fluoro-2-hydroxybenzene, 2-fluorohydroxybenzene, fluorophenol, unii-i9ow1nly9r, 2-fluoro-phenol, phenol, fluoro, ccris 1226
PubChem CID 9707
IUPAC Name 2-fluorophenol
SMILES OC1=CC=CC=C1F

Specifications

Color Yellow
Boiling Point 152°C
Quantity 25 g
UN Number 1993
Formula Weight 112.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluorophenol
RTECSNumber SL4530000
TSCA Yes