accessibility menu, dialog, popup

UserName

OHF

CAS RN 367-12-4

IUPAC Name: 2-fluorophenol
Synonyms: O-fluorophenol
Molecular Weight (g/mol): 112.1
Molecular Formula: C6H5FO
InChi Key: HFHFGHLXUCOHLN-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1F
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)