missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenol, 98%

CAS: 367-12-4 | C6H5FO | 112.10 g/mol

$87.06 - $87.06

Chemical Identifiers

CAS 367-12-4
Molecular Formula C6H5FO
Molecular Weight (g/mol) 112.10
MDL Number MFCD00002155
InChI Key HFHFGHLXUCOHLN-UHFFFAOYSA-N
Synonym o-fluorophenol, phenol, 2-fluoro, phenol, o-fluoro, 1-fluoro-2-hydroxybenzene, 2-fluorohydroxybenzene, fluorophenol, unii-i9ow1nly9r, 2-fluoro-phenol, phenol, fluoro, ccris 1226
PubChem CID 9707
IUPAC Name 2-fluorophenol
SMILES OC1=CC=CC=C1F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC119540100
SDS View Documents
Thermo Scientific Chemicals
119540100
10 g Glass bottle
Each for $87.06
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

367-12-4
112.10
HFHFGHLXUCOHLN-UHFFFAOYSA-N
9707
OC1=CC=CC=C1F
C6H5FO
MFCD00002155
o-fluorophenol, phenol, 2-fluoro, phenol, o-fluoro, 1-fluoro-2-hydroxybenzene, 2-fluorohydroxybenzene, fluorophenol, unii-i9ow1nly9r, 2-fluoro-phenol, phenol, fluoro, ccris 1226
2-fluorophenol

Specifications

367-12-4
100.0
Brown-Yellow to Colorless
152°C
Authentic
Glass bottle
1.5134 to 1.5154
MFCD00002155
06, I, 97
o-fluorophenol, phenol, 2-fluoro, phenol, o-fluoro, 1-fluoro-2-hydroxybenzene, 2-fluorohydroxybenzene, fluorophenol, unii-i9ow1nly9r, 2-fluoro-phenol, phenol, fluoro, ccris 1226
HFHFGHLXUCOHLN-UHFFFAOYSA-N
2-fluorophenol
9707
98%
2-Fluorophenol
97.5
16.1°C
1.2560g/mL
46°C
97.5% min. (GC)
C6H5FO
FC6H4OH
10 g
1.256
Solubility in water: 37.7g/L (20°C)
OC1=CC=CC=C1F
112.10
112.1
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

GHS Signal Word: Warning

EINECSNumber : 206-681-2

Recommended Storage : this product darkens during storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only