missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoronitrobenzene 98.0+%, TCI America™

Catalog No. p-7129496
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F010425G 25 g
F0104100G 100 g
F0104500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F010425G Supplier TCI America Supplier No. F010425G

Chemical Identifiers

CAS 1493-27-2
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.101
MDL Number MFCD00007048
InChI Key PWKNBLFSJAVFAB-UHFFFAOYSA-N
Synonym 2-fluoronitrobenzene, o-fluoronitrobenzene, o-nitrofluorobenzene, 2-nitrofluorobenzene, benzene, 1-fluoro-2-nitro, benzene, o-nitrofluoro, 2-fluoro-1-nitrobenzene, fluoronitrobenzene, 1-fluoro-2-nitro-benzene, benzene, fluoronitro
PubChem CID 73895
IUPAC Name 1-fluoro-2-nitrobenzene
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])F

Specifications

Color Brown-Yellow
Boiling Point 222°C
Quantity 25 g
UN Number 2810
Formula Weight 141.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoronitrobenzene
EINECSNumber (3)-0441
TSCA Yes