accessibility menu, dialog, popup

UserName

CAS RN 1493-27-2

IUPAC Name: 1-fluoro-2-nitrobenzene
Synonyms: O-fluoronitrobenzene
Molecular Weight (g/mol): 141.1
Molecular Formula: C6H4FNO2
InChi Key: PWKNBLFSJAVFAB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1F
Boiling Point 
  • (6)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (6)
  • (9)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)