missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoronitrobenzene 98.0+%, TCI America™
Supplier: TCI America F0104100G
Specifications
| 2-Fluoronitrobenzene | |
| Brown-Yellow | |
| C6H4FNO2 | |
| 100 g | |
| 2-fluoronitrobenzene, o-fluoronitrobenzene, o-nitrofluorobenzene, 2-nitrofluorobenzene, benzene, 1-fluoro-2-nitro, benzene, o-nitrofluoro, 2-fluoro-1-nitrobenzene, fluoronitrobenzene, 1-fluoro-2-nitro-benzene, benzene, fluoronitro | |
| C1=CC=C(C(=C1)[N+](=O)[O-])F | |
| 141.101 | |
| 141.10 | |
| Liquid |
| 1493-27-2 | |
| 222°C | |
| MFCD00007048 | |
| 2810 | |
| PWKNBLFSJAVFAB-UHFFFAOYSA-N | |
| 1-fluoro-2-nitrobenzene | |
| 73895 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1493-27-2 | |
| 141.101 | |
| PWKNBLFSJAVFAB-UHFFFAOYSA-N | |
| 73895 | |
| C1=CC=C(C(=C1)[N+](=O)[O-])F |
| C6H4FNO2 | |
| MFCD00007048 | |
| 2-fluoronitrobenzene, o-fluoronitrobenzene, o-nitrofluorobenzene, 2-nitrofluorobenzene, benzene, 1-fluoro-2-nitro, benzene, o-nitrofluoro, 2-fluoro-1-nitrobenzene, fluoronitrobenzene, 1-fluoro-2-nitro-benzene, benzene, fluoronitro | |
| 1-fluoro-2-nitrobenzene |
Safety and Handling
EINECSNumber : (3)-0441
TSCA : Yes