Learn More
2-Fluoromesitylene, 98%
CAS: 392-69-8 | C9H11F | 138.19 g/mol
Supplier: Thermo Scientific Chemicals A1277906
Description
2-Fluoromesitylene, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Fluoromesitylene, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents.
Specifications
| 2-Fluoromesitylene | |
| 0.975 | |
| C9H11F | |
| MFCD00052365 | |
| fluoromesitylene, mesityl fluoride, 2-fluoromesitylene, 2,4,6-trimethylfluorobenzene, benzene, 2-fluoro-1,3,5-trimethyl, 1-fluoro-2,4,6-trimethylbenzene, 2-fluoro-1,3,5-trimethyl-benzene, fluormesitylen, mesitylfluoride, pubchem16939 | |
| ZLGPNBBJPOBSLY-UHFFFAOYSA-N | |
| 2-fluoro-1,3,5-trimethylbenzene | |
| 136234 | |
| 98% |
| 392-69-8 | |
| 163°C to 165°C | |
| 1.483 | |
| 5 g | |
| Soluble in water. | |
| CC1=CC(C)=C(F)C(C)=C1 | |
| 138.19 | |
| 138.19 |
Chemical Identifiers
| 392-69-8 | |
| 138.19 | |
| ZLGPNBBJPOBSLY-UHFFFAOYSA-N | |
| 136234 | |
| CC1=CC(C)=C(F)C(C)=C1 |
| C9H11F | |
| MFCD00052365 | |
| fluoromesitylene, mesityl fluoride, 2-fluoromesitylene, 2,4,6-trimethylfluorobenzene, benzene, 2-fluoro-1,3,5-trimethyl, 1-fluoro-2,4,6-trimethylbenzene, 2-fluoro-1,3,5-trimethyl-benzene, fluormesitylen, mesitylfluoride, pubchem16939 | |
| 2-fluoro-1,3,5-trimethylbenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
RUO – Research Use Only