missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoromesitylene, 98%

Catalog No. AAA1277903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1277903 1 g
AAA1277906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1277903 Supplier Thermo Scientific Chemicals Supplier No. A1277903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 392-69-8 | C9H11F | 138.19 g/mol

2-Fluoromesitylene, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoromesitylene, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents.

Chemical Identifiers

CAS 392-69-8
Molecular Formula C9H11F
Molecular Weight (g/mol) 138.19
MDL Number MFCD00052365
InChI Key ZLGPNBBJPOBSLY-UHFFFAOYSA-N
Synonym fluoromesitylene, mesityl fluoride, 2-fluoromesitylene, 2,4,6-trimethylfluorobenzene, benzene, 2-fluoro-1,3,5-trimethyl, 1-fluoro-2,4,6-trimethylbenzene, 2-fluoro-1,3,5-trimethyl-benzene, fluormesitylen, mesitylfluoride, pubchem16939
PubChem CID 136234
IUPAC Name 2-fluoro-1,3,5-trimethylbenzene
SMILES CC1=CC(C)=C(F)C(C)=C1

Specifications

Density 0.975
Boiling Point 163°C to 165°C
Refractive Index 1.483
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 138.19
Percent Purity 98%
Chemical Name or Material 2-Fluoromesitylene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only