missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorofluorene 98.0+%, TCI America™

Catalog No. p-7129554
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02285G 5 g
F022825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02285G Supplier TCI America Supplier No. F02285G
Only null left

Chemical Identifiers

CAS 343-43-1
Molecular Formula C13H9F
Molecular Weight (g/mol) 184.213
MDL Number MFCD00031477
InChI Key SAKIZQTUUCRVJG-UHFFFAOYSA-N
PubChem CID 241132
IUPAC Name 2-fluoro-9H-fluorene
SMILES C1C2=CC=CC=C2C3=C1C=C(C=C3)F

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 5 g
Formula Weight 184.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluorofluorene