accessibility menu, dialog, popup

UserName

F

CAS RN 343-43-1

IUPAC Name: 2-fluoro-9H-fluorene
Synonyms: 2-fluoro-9H-fluorene
Molecular Weight (g/mol): 184.21
Molecular Formula: C13H9F
InChi Key: SAKIZQTUUCRVJG-UHFFFAOYSA-N
SMILES: FC1=CC=C2C(CC3=CC=CC=C23)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results