missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluorobenzyl alcohol, 98%
CAS: 446-51-5 | C7H7FO | 126.13 g/mol
$63.90 - $398.96
Chemical Identifiers
| CAS | 446-51-5 |
|---|---|
| Molecular Formula | C7H7FO |
| Molecular Weight (g/mol) | 126.13 |
| MDL Number | MFCD00004601 |
| InChI Key | QEHXDOJPVIHUDO-UHFFFAOYSA-N |
| Synonym | 2-fluorobenzyl alcohol, 2-fluorophenyl methanol, o-fluorobenzyl alcohol, benzenemethanol, 2-fluoro, 2-fluorobenzylalcohol, 2-fluoro-benzenemethanol, 2-fluorobenzylic alcohol, 2-fluorophenyl methan-1-ol, pubchem3485, acmc-1asq8 |
| PubChem CID | 67969 |
| IUPAC Name | (2-fluorophenyl)methanol |
| SMILES | OCC1=CC=CC=C1F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1479109
|
Thermo Scientific Chemicals
A1479109 |
10 g |
Each for $63.90
|
|
|||||
|
AAA1479118
|
Thermo Scientific Chemicals
A1479118 |
50 g |
Each for $243.62
|
|
|||||
|
AAA1479122
|
Thermo Scientific Chemicals
A1479122 |
100 g |
Each for $398.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 446-51-5 | |
| 126.13 | |
| QEHXDOJPVIHUDO-UHFFFAOYSA-N | |
| 67969 | |
| OCC1=CC=CC=C1F |
| C7H7FO | |
| MFCD00004601 | |
| 2-fluorobenzyl alcohol, 2-fluorophenyl methanol, o-fluorobenzyl alcohol, benzenemethanol, 2-fluoro, 2-fluorobenzylalcohol, 2-fluoro-benzenemethanol, 2-fluorobenzylic alcohol, 2-fluorophenyl methan-1-ol, pubchem3485, acmc-1asq8 | |
| (2-fluorophenyl)methanol |
Specifications
| 446-51-5 | |
| 201°C | |
| Characteristic | |
| 1.514 | |
| 10 g | |
| 2-fluorobenzyl alcohol, 2-fluorophenyl methanol, o-fluorobenzyl alcohol, benzenemethanol, 2-fluoro, 2-fluorobenzylalcohol, 2-fluoro-benzenemethanol, 2-fluorobenzylic alcohol, 2-fluorophenyl methan-1-ol, pubchem3485, acmc-1asq8 | |
| OCC1=CC=CC=C1F | |
| 126.13 | |
| 126.13 | |
| 2-Fluorobenzyl alcohol |
| 1.175 | |
| 90°C (194°F) | |
| C7H7FO | |
| MFCD00004601 | |
| 2079486 | |
| QEHXDOJPVIHUDO-UHFFFAOYSA-N | |
| (2-fluorophenyl)methanol | |
| 67969 | |
| 98% |
Safety and Handling
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
EINECSNumber : 207-170-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only