missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzyl alcohol, 98%

CAS: 446-51-5 | C7H7FO | 126.13 g/mol

Supplier:  Thermo Scientific Chemicals A1479118

 View more versions of this product

Catalog No. AAA1479118


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Fluorobenzyl alcohol
1.175
90°C (194°F)
C7H7FO
MFCD00004601
2079486
QEHXDOJPVIHUDO-UHFFFAOYSA-N
(2-fluorophenyl)methanol
67969
98%
446-51-5
201°C
Characteristic
1.514
50 g
2-fluorobenzyl alcohol, 2-fluorophenyl methanol, o-fluorobenzyl alcohol, benzenemethanol, 2-fluoro, 2-fluorobenzylalcohol, 2-fluoro-benzenemethanol, 2-fluorobenzylic alcohol, 2-fluorophenyl methan-1-ol, pubchem3485, acmc-1asq8
OCC1=CC=CC=C1F
126.13
126.13

Chemical Identifiers

446-51-5
126.13
QEHXDOJPVIHUDO-UHFFFAOYSA-N
67969
OCC1=CC=CC=C1F
C7H7FO
MFCD00004601
2-fluorobenzyl alcohol, 2-fluorophenyl methanol, o-fluorobenzyl alcohol, benzenemethanol, 2-fluoro, 2-fluorobenzylalcohol, 2-fluoro-benzenemethanol, 2-fluorobenzylic alcohol, 2-fluorophenyl methan-1-ol, pubchem3485, acmc-1asq8
(2-fluorophenyl)methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319

EINECSNumber : 207-170-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only