missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzotrifluoride 98.0+%, TCI America™

Supplier:  TCI America F038925G

 View more versions of this product

Catalog No. F038925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Fluorobenzotrifluoride
Yellow
C7H4F4
25 g
2-fluorobenzotrifluoride, 1-fluoro-2-trifluoromethyl benzene, o-fluorobenzotrifluoride, benzene, 1-fluoro-2-trifluoromethyl, alpha,alpha,alpha,2-tetrafluorotoluene, benzene, fluoro trifluoromethyl, o,alpha,alpha,alpha-tetrafluorotoluene, a,a,a,2-tetrafluorotoluene, alpha,alpha,alpha,ar-tetrafluorotoluene, 2-trifluoromethyl fluorobenzene
C1=CC=C(C(=C1)C(F)(F)F)F
164.103
164.10
Liquid
392-85-8
116°C
MFCD00000374
1993
BGVGHYOIWIALFF-UHFFFAOYSA-N
1-fluoro-2-(trifluoromethyl)benzene
9807
≥98.0% (GC)

Chemical Identifiers

392-85-8
164.103
BGVGHYOIWIALFF-UHFFFAOYSA-N
9807
C1=CC=C(C(=C1)C(F)(F)F)F
C7H4F4
MFCD00000374
2-fluorobenzotrifluoride, 1-fluoro-2-trifluoromethyl benzene, o-fluorobenzotrifluoride, benzene, 1-fluoro-2-trifluoromethyl, alpha,alpha,alpha,2-tetrafluorotoluene, benzene, fluoro trifluoromethyl, o,alpha,alpha,alpha-tetrafluorotoluene, a,a,a,2-tetrafluorotoluene, alpha,alpha,alpha,ar-tetrafluorotoluene, 2-trifluoromethyl fluorobenzene
1-fluoro-2-(trifluoromethyl)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions