missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7129656
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03895G 5 g
F038925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03895G Supplier TCI America Supplier No. F03895G
Only null left

Chemical Identifiers

CAS 392-85-8
Molecular Formula C7H4F4
Molecular Weight (g/mol) 164.103
MDL Number MFCD00000374
InChI Key BGVGHYOIWIALFF-UHFFFAOYSA-N
Synonym 2-fluorobenzotrifluoride, 1-fluoro-2-trifluoromethyl benzene, o-fluorobenzotrifluoride, benzene, 1-fluoro-2-trifluoromethyl, alpha,alpha,alpha,2-tetrafluorotoluene, benzene, fluoro trifluoromethyl, o,alpha,alpha,alpha-tetrafluorotoluene, a,a,a,2-tetrafluorotoluene, alpha,alpha,alpha,ar-tetrafluorotoluene, 2-trifluoromethyl fluorobenzene
PubChem CID 9807
IUPAC Name 1-fluoro-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)F

Specifications

Color Yellow
Boiling Point 116°C
Quantity 5 g
UN Number 1993
Formula Weight 164.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzotrifluoride