missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzophenone, 98+%

CAS: 342-24-5 | C13H9FO | 200.212 g/mol

Supplier:  Thermo Scientific Chemicals L0931614

 View more versions of this product

Catalog No. AAL0931614


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Fluorobenzophenone
1.186
>110°C (230°F)
1.5864
25 g
2-fluorobenzophenone, 2-fluorophenyl phenyl methanone, o-fluorobenzophenone, fluorobenzophenone, 2-fluorophenyl-phenylmethanone, 2-fluoro-phenyl-phenylmethanone, 2-fluorophenyl phenyl ketone, methanone, 2-fluorophenyl phenyl, pubchem4228, acmc-1ckt6
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2F
200.212
200.21
342-24-5
149°C to 150°C (16 mmHg)
C13H9FO
MFCD00000318
2047045
DWFDQVMFSLLMPE-UHFFFAOYSA-N
(2-fluorophenyl)-phenylmethanone
67650
≥98%

Chemical Identifiers

342-24-5
200.212
DWFDQVMFSLLMPE-UHFFFAOYSA-N
67650
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2F
C13H9FO
MFCD00000318
2-fluorobenzophenone, 2-fluorophenyl phenyl methanone, o-fluorobenzophenone, fluorobenzophenone, 2-fluorophenyl-phenylmethanone, 2-fluoro-phenyl-phenylmethanone, 2-fluorophenyl phenyl ketone, methanone, 2-fluorophenyl phenyl, pubchem4228, acmc-1ckt6
(2-fluorophenyl)-phenylmethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-440-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only