missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzophenone, 98+%

Catalog No. p-7045066
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0931606 5 g
AAL0931614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0931606 Supplier Thermo Scientific Chemicals Supplier No. L0931606
Only null left

CAS: 342-24-5 | C13H9FO | 200.212 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 342-24-5
Molecular Formula C13H9FO
Molecular Weight (g/mol) 200.212
MDL Number MFCD00000318
InChI Key DWFDQVMFSLLMPE-UHFFFAOYSA-N
Synonym 2-fluorobenzophenone, 2-fluorophenyl phenyl methanone, o-fluorobenzophenone, fluorobenzophenone, 2-fluorophenyl-phenylmethanone, 2-fluoro-phenyl-phenylmethanone, 2-fluorophenyl phenyl ketone, methanone, 2-fluorophenyl phenyl, pubchem4228, acmc-1ckt6
PubChem CID 67650
IUPAC Name (2-fluorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2F

Specifications

Density 1.186
Boiling Point 149°C to 150°C (16 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5864
Quantity 5 g
Beilstein 2047045
Formula Weight 200.21
Percent Purity ≥98%
Chemical Name or Material 2-Fluorobenzophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-440-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only