Learn More
2-Fluorobenzeneboronic acid, 98%
CAS: 1993-03-9 | C6H6BFO2 | 139.92 g/mol
$82.26 - $813.81
Chemical Identifiers
| CAS | 1993-03-9 |
|---|---|
| Molecular Formula | C6H6BFO2 |
| Molecular Weight (g/mol) | 139.92 |
| MDL Number | MFCD00674013 |
| InChI Key | QCSLIRFWJPOENV-UHFFFAOYSA-N |
| Synonym | 2-fluorophenyl boronic acid, 2-fluorobenzeneboronic acid, 2-fluorophenyl boranediol, o-fluorophenylboronic acid, boronic acid, b-2-fluorophenyl, boronic acid, 2-fluorophenyl, 2-fluoro-phenyl-boronic acid, o-fluoro-benzeneboronic acid, benzeneboronic acid, o-fluoro |
| PubChem CID | 2734354 |
| IUPAC Name | (2-fluorophenyl)boronic acid |
| SMILES | OB(O)C1=CC=CC=C1F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2310303
|
Thermo Scientific Chemicals
B2310303 |
1 g |
Each for $82.26
|
|
|||||
|
AAB2310306
|
Thermo Scientific Chemicals
B2310306 |
5 g |
Each for $201.25
|
|
|||||
|
AAB2310314
|
Thermo Scientific Chemicals
B2310314 |
25 g |
Each for $813.81
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1993-03-9 | |
| 139.92 | |
| QCSLIRFWJPOENV-UHFFFAOYSA-N | |
| 2734354 | |
| OB(O)C1=CC=CC=C1F |
| C6H6BFO2 | |
| MFCD00674013 | |
| 2-fluorophenyl boronic acid, 2-fluorobenzeneboronic acid, 2-fluorophenyl boranediol, o-fluorophenylboronic acid, boronic acid, b-2-fluorophenyl, boronic acid, 2-fluorophenyl, 2-fluoro-phenyl-boronic acid, o-fluoro-benzeneboronic acid, benzeneboronic acid, o-fluoro | |
| (2-fluorophenyl)boronic acid |
Specifications
| 1993-03-9 | |
| C6H6BFO2 | |
| 1 g | |
| 2-fluorophenyl boronic acid, 2-fluorobenzeneboronic acid, 2-fluorophenyl boranediol, o-fluorophenylboronic acid, boronic acid, b-2-fluorophenyl, boronic acid, 2-fluorophenyl, 2-fluoro-phenyl-boronic acid, o-fluoro-benzeneboronic acid, benzeneboronic acid, o-fluoro | |
| OB(O)C1=CC=CC=C1F | |
| 139.92 | |
| 139.92 | |
| 2-Fluorobenzeneboronic acid |
| 100°C to 108°C | |
| MFCD00674013 | |
| 3030413 | |
| QCSLIRFWJPOENV-UHFFFAOYSA-N | |
| (2-fluorophenyl)boronic acid | |
| 2734354 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only