accessibility menu, dialog, popup

UserName

CAS RN 1993-03-9

IUPAC Name: (2-fluorophenyl)boronic acid
Synonyms: 2-fluorophenylboronic acid
Molecular Weight (g/mol): 139.92
Molecular Formula: C6H6BFO2
InChi Key: QCSLIRFWJPOENV-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1F
Quantity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (3)