missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzeneboronic acid, 98%

Catalog No. AAB2310303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2310303 1 g
AAB2310306 5 g
AAB2310314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2310303 Supplier Thermo Scientific Chemicals Supplier No. B2310303

CAS: 1993-03-9 | C6H6BFO2 | 139.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1993-03-9
Molecular Formula C6H6BFO2
Molecular Weight (g/mol) 139.92
MDL Number MFCD00674013
InChI Key QCSLIRFWJPOENV-UHFFFAOYSA-N
Synonym 2-fluorophenyl boronic acid, 2-fluorobenzeneboronic acid, 2-fluorophenyl boranediol, o-fluorophenylboronic acid, boronic acid, b-2-fluorophenyl, boronic acid, 2-fluorophenyl, 2-fluoro-phenyl-boronic acid, o-fluoro-benzeneboronic acid, benzeneboronic acid, o-fluoro
PubChem CID 2734354
IUPAC Name (2-fluorophenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1F

Specifications

Melting Point 100°C to 108°C
Quantity 1 g
Beilstein 3030413
Formula Weight 139.92
Percent Purity 98%
Chemical Name or Material 2-Fluorobenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only