missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzaldehyde 98.0+%, TCI America™

Catalog No. F019025ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F019025ML 25 mL
F0190250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F019025ML Supplier TCI America Supplier No. F019025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 446-52-6
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.114
MDL Number MFCD00003302
InChI Key ZWDVQMVZZYIAHO-UHFFFAOYSA-N
Synonym o-fluorobenzaldehyde, benzaldehyde, 2-fluoro, benzaldehyde, o-fluoro, unii-7zx20dxb9z, 2-fluor-benzaldehyd, 2-fluoro-benzaldehyde, benzaldehyde, fluoro, ortho-fluorobenzaldehyde, 7zx20dxb9z, fluorobenzaldehyde
PubChem CID 67970
IUPAC Name 2-fluorobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)F

Specifications

Melting Point -45°C
Color Yellow
Boiling Point 175°C
Quantity 25 mL
UN Number 1989
Formula Weight 124.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzaldehyde
RTECSNumber CU6140000
TSCA No