accessibility menu, dialog, popup

UserName

FO

CAS RN 446-52-6

IUPAC Name: 2-fluorobenzaldehyde
Synonyms: 2-fluor-benzaldehyd
Molecular Weight (g/mol): 124.11
Molecular Formula: C7H5FO
InChi Key: ZWDVQMVZZYIAHO-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C=O
Boiling Point 
  • (6)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)