missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-p-anisaldehyde 96.0+%, TCI America™
Supplier: TCI America F038625G
Specifications
| 2-Fluoro-p-anisaldehyde | |
| 46°C | |
| MFCD00236679 | |
| 2-fluoro-p-anisaldehyde, 2-fluoro-4-methoxy-benzaldehyde, 2-fluoro-4-methoxy benzaldehyde, benzaldehyde, 2-fluoro-4-methoxy, 2-fluoro-4-methoxybenzaldhyde, pubchem1447, acmc-209hyt, 3-fluoro-4-formylanisole, 2-fluor-4-methoxybenzaldehyd | |
| COC1=CC=C(C=O)C(F)=C1 | |
| 154.14 | |
| 154.14 | |
| Crystalline Powder |
| 331-64-6 | |
| C8H7FO2 | |
| 25 g | |
| UNWQNFJBBWXFBG-UHFFFAOYSA-N | |
| 2-fluoro-4-methoxybenzaldehyde | |
| 2734873 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 331-64-6 | |
| 154.14 | |
| UNWQNFJBBWXFBG-UHFFFAOYSA-N | |
| 2734873 | |
| COC1=CC=C(C=O)C(F)=C1 |
| C8H7FO2 | |
| MFCD00236679 | |
| 2-fluoro-p-anisaldehyde, 2-fluoro-4-methoxy-benzaldehyde, 2-fluoro-4-methoxy benzaldehyde, benzaldehyde, 2-fluoro-4-methoxy, 2-fluoro-4-methoxybenzaldhyde, pubchem1447, acmc-209hyt, 3-fluoro-4-formylanisole, 2-fluor-4-methoxybenzaldehyd | |
| 2-fluoro-4-methoxybenzaldehyde |
Safety and Handling
TSCA : No