missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-p-anisaldehyde 96.0+%, TCI America™

Catalog No. p-7129654
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03865G 5 g
F038625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03865G Supplier TCI America Supplier No. F03865G
Only null left

Chemical Identifiers

CAS 331-64-6
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00236679
InChI Key UNWQNFJBBWXFBG-UHFFFAOYSA-N
Synonym 2-fluoro-p-anisaldehyde, 2-fluoro-4-methoxy-benzaldehyde, 2-fluoro-4-methoxy benzaldehyde, benzaldehyde, 2-fluoro-4-methoxy, 2-fluoro-4-methoxybenzaldhyde, pubchem1447, acmc-209hyt, 3-fluoro-4-formylanisole, 2-fluor-4-methoxybenzaldehyd
PubChem CID 2734873
IUPAC Name 2-fluoro-4-methoxybenzaldehyde
SMILES COC1=CC=C(C=O)C(F)=C1

Specifications

Melting Point 46°C
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-p-anisaldehyde