missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-N-methylbenzylamine, 95%

Catalog No. p-7052628
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5443903 1 g
AAH5443906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5443903 Supplier Thermo Scientific Chemicals Supplier No. H5443903
Only null left

CAS: 399-30-4 | C8H10FN | 139.173 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and air. Store at 4-2°C.

Chemical Identifiers

CAS 399-30-4
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD04629633
InChI Key AHIHZCXUWGORQO-UHFFFAOYSA-N
Synonym n-methyl-2-fluorobenzylamine, 2-fluorobenzyl methylamine, 2-fluoro-n-methylbenzylamine, 2-fluorophenyl methyl methyl amine, 1-2-fluorophenyl-n-methylmethanamine, n-2-fluorobenzyl-n-methylamine, n-methyl-o-fluorobenzylamine, benzenemethanamine, 2-fluoro-n-methyl, 1-2-fluorophenyl-n-methyl-methanamine, acmc-209zio
PubChem CID 2060842
IUPAC Name 1-(2-fluorophenyl)-N-methylmethanamine
SMILES CNCC1=CC=CC=C1F

Specifications

Color Colorless to Tan
Density 1.047 g/mL
Boiling Point 73°C
Flash Point 68°C (154°F)
Refractive Index 1.047
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 139.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-N-methylbenzylamine
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only