CAS RN 399-30-4
IUPAC Name:
[(2-fluorophenyl)methyl](methyl)azanium
Synonyms:
[(2-fluorophenyl)methyl](methyl)azanium
Molecular Weight (g/mol):
140.18
Molecular Formula:
C8H11FN
InChi Key:
AHIHZCXUWGORQO-UHFFFAOYSA-O
SMILES:
C[NH2+]CC1=CC=CC=C1F
1
–
1
of
1
results
1
Promotions Available
2-Fluoro-N-methylbenzylamine, 95%
2-Fluoro-N-methylbenzylamine, 95%
CAS: 399-30-4 Molecular Formula: C8H10FN Molecular Weight (g/mol): 139.173 MDL Number: MFCD04629633 InChI Key: AHIHZCXUWGORQO-UHFFFAOYSA-N Synonym: n-methyl-2-fluorobenzylamine,2-fluorobenzyl methylamine,2-fluoro-n-methylbenzylamine,2-fluorophenyl methyl methyl amine,1-2-fluorophenyl-n-methylmethanamine,n-2-fluorobenzyl-n-methylamine,n-methyl-o-fluorobenzylamine,benzenemethanamine, 2-fluoro-n-methyl,1-2-fluorophenyl-n-methyl-methanamine,acmc-209zio PubChem CID: 2060842 IUPAC Name: 1-(2-fluorophenyl)-N-methylmethanamine SMILES: CNCC1=CC=CC=C1F