missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-N-methylbenzamide, 97%

Catalog No. p-7037383
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56104MD 250 mg
AAH5610403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH56104MD Supplier Thermo Scientific Chemicals Supplier No. H56104MD

CAS: 52833-63-3 | C8H8FNO | 153.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52833-63-3
Molecular Formula C8H8FNO
Molecular Weight (g/mol) 153.16
MDL Number MFCD01214098
InChI Key NAGFMACWWJYORB-UHFFFAOYSA-N
PubChem CID 2425972
IUPAC Name 2-fluoro-N-methylbenzamide
SMILES CNC(=O)C1=CC=CC=C1F

Specifications

Quantity 250 mg
Synonym o-fluoro-n-methylbenzamide, 2-fluoro-n-methyl-benzamide, benzamide,2-fluoro-n-methyl, benzamide, 2-fluoro-n-methyl, 2-fluorophenyl-n-methylcarboxamide
Formula Weight 153.16
Percent Purity 97%
Chemical Name or Material 2-Fluoro-N-methylbenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only