accessibility menu, dialog, popup

UserName

CAS RN 52833-63-3

IUPAC Name: 2-fluoro-N-methylbenzamide
Synonyms: 2-fluoro-N-methylbenzamide
Molecular Weight (g/mol): 153.16
Molecular Formula: C8H8FNO
InChi Key: NAGFMACWWJYORB-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results