missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-m-xylene 98.0+%, TCI America™

Catalog No. p-7129665
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
F04025G 5 g
F040225G 25 g
2 options

Catalog No. F04025G

Supplier: TCI America F04025G

Only null left

Chemical Identifiers

CAS 443-88-9
Molecular Formula C8H9F
Molecular Weight (g/mol) 124.158
MDL Number MFCD00039215
InChI Key JTAUTNBVFDTYTI-UHFFFAOYSA-N
Synonym 2,6-dimethylfluorobenzene, 2-fluoro-m-xylene, 1-fluoro-2,6-dimethylbenzene, 1,3-dimethyl-2-fluorobenzene, benzene, 2-fluoro-1,3-dimethyl, 2,6-dimethyl fluorobenzene, 2-fluoro-1,3-dimethyl-benzene, fr b1 f1, pubchem4407, 2-fluoro-3-methyltoluene
PubChem CID 123065
IUPAC Name 2-fluoro-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)F

Specifications

Color Yellow
Boiling Point 167°C
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 1993
Formula Weight 124.16
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Fluoro-m-xylene