accessibility menu, dialog, popup

UserName

CH3H3CF

CAS RN 443-88-9

IUPAC Name: 2-fluoro-1,3-dimethylbenzene
Synonyms: 2-fluoro-1,3-dimethylbenzene
Molecular Weight (g/mol): 124.16
Molecular Formula: C8H9F
InChi Key: JTAUTNBVFDTYTI-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1F
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)