missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-DL-mandelic Acid 98.0+%, TCI America™

Catalog No. F07305G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07305G 5 g
F073025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07305G Supplier TCI America Supplier No. F07305G
Only null left

Chemical Identifiers

CAS 389-31-1
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.14
MDL Number MFCD03095382
InChI Key WWSRHIZZWWDONI-UHFFFAOYNA-N
Synonym 2-fluoromandelic acid, 2-2-fluorophenyl-2-hydroxyacetic acid, 2-fluorophenyl hydroxy acetic acid, o-fluoromandelic acid, 2-fluoro-dl-mandelic acid, 2-fluorophenyl hydroxyacetic acid, 2-fluoro-alpha-hydroxyphenylacetic acid, 2-2-fluorophenyl-2-hydroxyaceticacid, benzeneacetic acid,2-fluoro-a-hydroxy-, ar
PubChem CID 2783169
IUPAC Name 2-(2-fluorophenyl)-2-hydroxyacetic acid
SMILES OC(C(O)=O)C1=CC=CC=C1F

Specifications

Melting Point 117°C
Color White-Yellow
Boiling Point 72°C
Quantity 5 g
Formula Weight 170.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-DL-mandelic Acid