accessibility menu, dialog, popup

UserName

CAS RN 389-31-1

OOHHOF

CAS RN 389-31-1

IUPAC Name: 2-(2-fluorophenyl)-2-hydroxyacetic acid
Synonyms: (2-fluorophenyl)(hydroxy)acetic acid
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: WWSRHIZZWWDONI-UHFFFAOYNA-N
SMILES: OC(C(O)=O)C1=CC=CC=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results