missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-6-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™
$73.16 - $221.39
Chemical Identifiers
| CAS | 32890-94-1 |
|---|---|
| Molecular Formula | C8H3F4O2 |
| Molecular Weight (g/mol) | 207.10 |
| MDL Number | MFCD00040981 |
| InChI Key | LNARMXLVVGHCRP-UHFFFAOYSA-M |
| Synonym | 2-fluoro-6-trifluoromethyl benzoic acid, otf-boa-6f, alpha,alpha,alpha,6-tetrafluoro-o-toluic acid, benzoic acid, 2-fluoro-6-trifluoromethyl, 6-fluoro-2-trifluoromethylbenzoic acid, 2-fluoro-6-trifluoromethyl-benzoic acid, 6-fluoro-2-trifluoromethyl benzoic acid, pubchem4759, acmc-20a40d, rarechem al bo 0626 |
| PubChem CID | 688123 |
| IUPAC Name | 2-fluoro-6-(trifluoromethyl)benzoate |
| SMILES | [O-]C(=O)C1=C(F)C=CC=C1C(F)(F)F |
Chemical Identifiers
| 32890-94-1 | |
| 207.10 | |
| LNARMXLVVGHCRP-UHFFFAOYSA-M | |
| 688123 | |
| [O-]C(=O)C1=C(F)C=CC=C1C(F)(F)F |
| C8H3F4O2 | |
| MFCD00040981 | |
| 2-fluoro-6-trifluoromethyl benzoic acid, otf-boa-6f, alpha,alpha,alpha,6-tetrafluoro-o-toluic acid, benzoic acid, 2-fluoro-6-trifluoromethyl, 6-fluoro-2-trifluoromethylbenzoic acid, 2-fluoro-6-trifluoromethyl-benzoic acid, 6-fluoro-2-trifluoromethyl benzoic acid, pubchem4759, acmc-20a40d, rarechem al bo 0626 | |
| 2-fluoro-6-(trifluoromethyl)benzoate |
Specifications
| 32890-94-1 | |
| White | |
| MFCD00040981 | |
| 2-fluoro-6-trifluoromethyl benzoic acid, otf-boa-6f, alpha,alpha,alpha,6-tetrafluoro-o-toluic acid, benzoic acid, 2-fluoro-6-trifluoromethyl, 6-fluoro-2-trifluoromethylbenzoic acid, 2-fluoro-6-trifluoromethyl-benzoic acid, 6-fluoro-2-trifluoromethyl benzoic acid, pubchem4759, acmc-20a40d, rarechem al bo 0626 | |
| [O-]C(=O)C1=C(F)C=CC=C1C(F)(F)F | |
| 207.10 | |
| 208.11 | |
| Crystalline Powder |
| 88°C | |
| C8H3F4O2 | |
| 1 g | |
| LNARMXLVVGHCRP-UHFFFAOYSA-M | |
| 2-fluoro-6-(trifluoromethyl)benzoate | |
| 688123 | |
| ≥98.0% (GC,T) | |
| 2-Fluoro-6-(trifluoromethyl)benzoic Acid |
Safety and Handling
TSCA : No