missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-6-methylaniline 98.0+%, TCI America™
$46.60 - $126.76
Chemical Identifiers
| CAS | 443-89-0 |
|---|---|
| Molecular Formula | C7H8FN |
| Molecular Weight (g/mol) | 125.146 |
| MDL Number | MFCD06658252 |
| InChI Key | CMVJYZNBMRJICR-UHFFFAOYSA-N |
| Synonym | 2-amino-3-fluorotoluene, 3-fluoro-2-aminotoluene, 6-fluoro-o-toluidine, 2-fluoro-6-methyl-phenylamine, 2-fluoro-6-methylbenzenamine, 6-fluoro-2-methylphenylamine, benzenamine, 2-fluoro-6-methyl, pubchem4470, acmc-209jxr, 2-fluoro-6-methyl aniline |
| PubChem CID | 14155411 |
| IUPAC Name | 2-fluoro-6-methylaniline |
| SMILES | CC1=C(C(=CC=C1)F)N |
Chemical Identifiers
| 443-89-0 | |
| 125.146 | |
| CMVJYZNBMRJICR-UHFFFAOYSA-N | |
| 14155411 | |
| CC1=C(C(=CC=C1)F)N |
| C7H8FN | |
| MFCD06658252 | |
| 2-amino-3-fluorotoluene, 3-fluoro-2-aminotoluene, 6-fluoro-o-toluidine, 2-fluoro-6-methyl-phenylamine, 2-fluoro-6-methylbenzenamine, 6-fluoro-2-methylphenylamine, benzenamine, 2-fluoro-6-methyl, pubchem4470, acmc-209jxr, 2-fluoro-6-methyl aniline | |
| 2-fluoro-6-methylaniline |
Specifications
| 443-89-0 | |
| 89°C | |
| MFCD06658252 | |
| 2-amino-3-fluorotoluene, 3-fluoro-2-aminotoluene, 6-fluoro-o-toluidine, 2-fluoro-6-methyl-phenylamine, 2-fluoro-6-methylbenzenamine, 6-fluoro-2-methylphenylamine, benzenamine, 2-fluoro-6-methyl, pubchem4470, acmc-209jxr, 2-fluoro-6-methyl aniline | |
| CC1=C(C(=CC=C1)F)N | |
| 125.146 | |
| 125.15 | |
| Liquid |
| Yellow | |
| C7H8FN | |
| 1 g | |
| CMVJYZNBMRJICR-UHFFFAOYSA-N | |
| 2-fluoro-6-methylaniline | |
| 14155411 | |
| ≥98.0% (T) | |
| 2-Fluoro-6-methylaniline |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator