accessibility menu, dialog, popup

UserName

CAS RN 443-89-0

CH3FNH2

CAS RN 443-89-0

IUPAC Name: 2-fluoro-6-methylaniline
Synonyms: 2-fluoro-6-methylaniline
Molecular Weight (g/mol): 125.15
Molecular Formula: C7H8FN
InChi Key: CMVJYZNBMRJICR-UHFFFAOYSA-N
SMILES: CC1=C(N)C(F)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results