missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-6-methylaniline 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 89°C |
| Quantity | 5 g |
| Formula Weight | 125.15 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Fluoro-6-methylaniline |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |