missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methylaniline 98.0+%, TCI America™

Catalog No. F08171G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08171G 1 g
F08175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08171G Supplier TCI America Supplier No. F08171G
Only null left

Chemical Identifiers

CAS 443-89-0
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
MDL Number MFCD06658252
InChI Key CMVJYZNBMRJICR-UHFFFAOYSA-N
Synonym 2-amino-3-fluorotoluene, 3-fluoro-2-aminotoluene, 6-fluoro-o-toluidine, 2-fluoro-6-methyl-phenylamine, 2-fluoro-6-methylbenzenamine, 6-fluoro-2-methylphenylamine, benzenamine, 2-fluoro-6-methyl, pubchem4470, acmc-209jxr, 2-fluoro-6-methyl aniline
PubChem CID 14155411
IUPAC Name 2-fluoro-6-methylaniline
SMILES CC1=C(C(=CC=C1)F)N

Specifications

Color Yellow
Boiling Point 89°C
Quantity 1 g
Formula Weight 125.15
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-6-methylaniline
TSCA No
Recommended Storage Refrigerator