missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxyphenol, 97%

Catalog No. p-7029039
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2647103 1 g
AAH2647106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2647103 Supplier Thermo Scientific Chemicals Supplier No. H2647103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 73943-41-6 | C7H7FO2 | 142.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73943-41-6
Molecular Formula C7H7FO2
Molecular Weight (g/mol) 142.13
MDL Number MFCD00075140
InChI Key YZNHPLVFLRSVHY-UHFFFAOYSA-N
Synonym 2-fluoro-6-methoxy-phenol, phenol, 2-fluoro-6-methoxy, pubchem3878, acmc-209osz, 2-methoxy-6-fluorophenol, 2-methoxyl-6-fluorophenol, 2-hydroxy-3-fluoroanisole, 2-fluoro-6-methoxy phenol, phenol,2-fluoro-6-methoxy, 2-fluoro-6-methoxyphenol
PubChem CID 2737367
IUPAC Name 2-fluoro-6-methoxyphenol
SMILES COC1=C(O)C(F)=CC=C1

Specifications

Density 1.23
Boiling Point 130°C to 131°C (36 mmHg)
Flash Point 103°C (218°F)
Refractive Index 1.52
Quantity 1 g
Formula Weight 142.13
Percent Purity 97%
Chemical Name or Material 2-Fluoro-6-methoxyphenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only