accessibility menu, dialog, popup

UserName

CAS RN 73943-41-6

IUPAC Name: 2-fluoro-6-methoxyphenol
Synonyms: 2-fluoro-6-methoxyphenol
Molecular Weight (g/mol): 142.13
Molecular Formula: C7H7FO2
InChi Key: YZNHPLVFLRSVHY-UHFFFAOYSA-N
SMILES: COC1=C(O)C(F)=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)