missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxybenzyl bromide, 98%

Catalog No. AAH26136MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26136MD 250 mg
AAH2613603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH26136MD Supplier Thermo Scientific Chemicals Supplier No. H26136MD

CAS: 500912-17-4 | C8H8BrFO | 219.053 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 500912-17-4
Molecular Formula C8H8BrFO
Molecular Weight (g/mol) 219.053
MDL Number MFCD08688579
InChI Key PVKFBCBTHHTDEX-UHFFFAOYSA-N
Synonym 2-fluoro-6-methoxybenzyl bromide, 2-bromomethyl-1-fluoro-3-methoxybenzene, 2-bromomethyl-3-fluoroanisole, 2-fluoro-6-methoxy-benzyl bromide, 2-bromomethyl-3-fluorophenyl methyl ether, benzene,2-bromomethyl-1-fluoro-3-methoxy, 2-bromomethyl-1-fluoranyl-3-methoxy-benzene
PubChem CID 2779322
IUPAC Name 2-(bromomethyl)-1-fluoro-3-methoxybenzene
SMILES COC1=C(C(=CC=C1)F)CBr

Specifications

Melting Point 18°C to 21°C
Refractive Index 1.557
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 219.05
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-methoxybenzyl bromide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only