accessibility menu, dialog, popup

UserName

H3COFBr

CAS RN 500912-17-4

IUPAC Name: 2-(bromomethyl)-1-fluoro-3-methoxybenzene
Synonyms: 2-(bromomethyl)-1-fluoro-3-methoxybenzene
Molecular Weight (g/mol): 219.05
Molecular Formula: C8H8BrFO
InChi Key: PVKFBCBTHHTDEX-UHFFFAOYSA-N
SMILES: COC1=C(CBr)C(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results