missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-iodotoluene 98.0+%, TCI America™

Catalog No. F11345G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F11345G Supplier TCI America Supplier No. F11345G
Only null left

Chemical Identifiers

CAS 443-85-6
Molecular Formula C7H6FI
Molecular Weight (g/mol) 236.03
MDL Number MFCD00142922
InChI Key MSPXWJMFEVAKHQ-UHFFFAOYSA-N
Synonym 2-fluoro-6-iodotoluene, pubchem1620, acmc-209xbg, ksc493m7j, benzene, 1-fluoro-3-iodo-2-methyl, 1-fluoranyl-3-iodanyl-2-methyl-benzene
PubChem CID 2774523
IUPAC Name 1-fluoro-3-iodo-2-methylbenzene
SMILES CC1=C(F)C=CC=C1I

Specifications

Color Yellow
Boiling Point 117°C
Quantity 5 g
Formula Weight 236.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-6-iodotoluene