CAS RN 443-85-6
IUPAC Name:
1-fluoro-3-iodo-2-methylbenzene
Synonyms:
1-fluoro-3-iodo-2-methylbenzene
Molecular Weight (g/mol):
236.03
Molecular Formula:
C7H6FI
InChi Key:
MSPXWJMFEVAKHQ-UHFFFAOYSA-N
SMILES:
CC1=C(F)C=CC=C1I
1
–
1
of
1
results
1
2-Fluoro-6-iodotoluene 98.0+%, TCI America™
CAS: 443-85-6 Molecular Formula: C7H6FI Molecular Weight (g/mol): 236.03 MDL Number: MFCD00142922 InChI Key: MSPXWJMFEVAKHQ-UHFFFAOYSA-N Synonym: 2-fluoro-6-iodotoluene,pubchem1620,acmc-209xbg,ksc493m7j,benzene, 1-fluoro-3-iodo-2-methyl,1-fluoranyl-3-iodanyl-2-methyl-benzene PubChem CID: 2774523 IUPAC Name: 1-fluoro-3-iodo-2-methylbenzene SMILES: CC1=C(F)C=CC=C1I