missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-hydroxybenzonitrile 98.0+%, TCI America™

Catalog No. F09851G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09851G 1 g
F09855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09851G Supplier TCI America Supplier No. F09851G

Chemical Identifiers

CAS 140675-43-0
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.113
MDL Number MFCD03428592
InChI Key YEBHNFDMNFHZFF-UHFFFAOYSA-N
Synonym 2-cyano-3-fluorophenol, 6-fluorosalicylonitrile, 2-fluoro-6-hydroxy-benzonitrile, 6-fluoro-2-hydroxybenzonitrile, benzonitrile, 2-fluoro-6-hydroxy, 2-fluoro-6-hydroxybenzonitrle, pubchem2223, acmc-1c03f, 2-fuoro-6-hydroxybenzonitrle
PubChem CID 2783216
IUPAC Name 2-fluoro-6-hydroxybenzonitrile
SMILES C1=CC(=C(C(=C1)F)C#N)O

Specifications

Melting Point 157°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 137.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-6-hydroxybenzonitrile