missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™

Catalog No. p-7129790
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06201G 1 g
F06205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06201G Supplier TCI America Supplier No. F06201G

Chemical Identifiers

CAS 115029-23-7
Molecular Formula C8H3F4O2
Molecular Weight (g/mol) 207.10
MDL Number MFCD00061292
InChI Key LIFKXWNFWIUMJT-UHFFFAOYSA-M
Synonym 2-fluoro-5-trifluoromethyl benzoic acid, 2-fluoro-5-trifluoromethyl benzoicacid, alpha,alpha,alpha,6-tetrafluoro-m-toluic acid, benzoic acid, 2-fluoro-5-trifluoromethyl, maybridge1_006396, 2-fluoro-5-trifluoromethylbenzioc acid, pubchem4758, acmc-1bvsf, ksc180k3f
PubChem CID 688292
IUPAC Name 2-fluoro-5-(trifluoromethyl)benzoate
SMILES [O-]C(=O)C1=CC(=CC=C1F)C(F)(F)F

Specifications

Melting Point 100°C
Color Yellow
Quantity 1 g
Formula Weight 208.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzoic Acid