missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)anisole, 97%

Catalog No. AAB2351603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2351603 1 g
AAB2351606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2351603 Supplier Thermo Scientific Chemicals Supplier No. B2351603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 261951-78-4 | C8H6F4O | 194.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Chemical Identifiers

CAS 261951-78-4
Molecular Formula C8H6F4O
Molecular Weight (g/mol) 194.129
MDL Number MFCD01631564
InChI Key AXEAQZIYCQTHQV-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethyl anisole, 1-fluoro-2-methoxy-4-trifluoromethyl benzene, 4-fluoro-3-methoxybenzotrifluoride, pubchem8275, acmc-1ceet, 4-fluoro-3-methoxy-benzotrifluoride, 1-fluoranyl-2-methoxy-4-trifluoromethyl benzene
PubChem CID 2774771
IUPAC Name 1-fluoro-2-methoxy-4-(trifluoromethyl)benzene
SMILES COC1=C(C=CC(=C1)C(F)(F)F)F

Specifications

Flash Point 62°C (144°F)
Refractive Index 1.4352
Quantity 1 g
Solubility Information Not miscible in water.
Formula Weight 194.13
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)anisole
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only