missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)aniline, 97%

Catalog No. p-7023721
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1755603 1 g
AAA1755606 5 g
AAA1755614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1755603 Supplier Thermo Scientific Chemicals Supplier No. A1755603

CAS: 535-52-4 | C7H5F4N | 179.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 535-52-4
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.118
MDL Number MFCD00007653
InChI Key DRKWGMXFFCPZLW-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethyl aniline, 3-amino-4-fluorobenzotrifluoride, alpha,alpha,alpha,6-tetrafluoro-m-toluidine, 4-fluoro-3-aminobenzotrifluoride, 2-fluoro-5-trifluoromethyl benzenamine, 2-fluoro-5-trifluoromethyl phenylamine, benzenamine, 2-fluoro-5-trifluoromethyl, .alpha.,.alpha.,.alpha.,6-tetrafluoro-m-toluidine, pubchem1538, ksc494s7l
PubChem CID 136349
IUPAC Name 2-fluoro-5-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1C(F)(F)F)N)F

Specifications

Density 1.378
Boiling Point 210°C
Flash Point 70°C (158°F)
Odor Characteristic
Refractive Index 1.461
Quantity 1 g
UN Number UN2810
Beilstein 1950800
Formula Weight 179.12
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)aniline
Show More Show Less
Hazard Category H227-H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H227-H302-H332-H315-H319
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only